Learn More
2,3-Dichlorobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals
$472.71 - $472.71
Chemical Identifiers
CAS | 82417-45-6 |
---|---|
Molecular Formula | C6H3Cl3O2S |
Molecular Weight (g/mol) | 245.498 |
MDL Number | MFCD00051844 |
InChI Key | PQODWTNHDKDHIW-UHFFFAOYSA-N |
Synonym | 2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r |
PubChem CID | 2734222 |
IUPAC Name | 2,3-dichlorobenzenesulfonyl chloride |
SMILES | C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1228314
|
Thermo Scientific Chemicals
A1228314 |
25 g |
Each for $472.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
82417-45-6 | |
245.498 | |
PQODWTNHDKDHIW-UHFFFAOYSA-N | |
2734222 | |
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl |
C6H3Cl3O2S | |
MFCD00051844 | |
2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r | |
2,3-dichlorobenzenesulfonyl chloride |
Specifications
82417-45-6 | |
131°C to 133°C (1 mmHg) | |
MFCD00051844 | |
UN3261 | |
Moisture sensitive | |
PQODWTNHDKDHIW-UHFFFAOYSA-N | |
2,3-dichlorobenzenesulfonyl chloride | |
2734222 | |
98% |
57°C to 62°C | |
C6H3Cl3O2S | |
25 g | |
7703966 | |
2,3-dichlorobenzene-1-sulfonyl chloride, 2,3-dichlorobenzenesulphonyl chloride, 2,3-dichloro-benzenesulfonyl chloride, 2,3-dichlorobenzenesulphonylchloride, 2,3-dichlorophenylsulfonyl chloride, benzenesulfonyl chloride, 2,3-dichloro-9ci, 2,3-dichlorobenzenesulfonylchloride, pubchem5114, buttpark 9357-48, acmc-1bl9r | |
C1=CC(=C(C(=C1)Cl)Cl)S(=O)(=O)Cl | |
245.498 | |
245.51 | |
2,3-Dichlorobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only