missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorobenzoyl Chloride 98.0+%, TCI America™
$70.84 - $677.78
Chemical Identifiers
| CAS | 2905-60-4 |
|---|---|
| Molecular Formula | C7H3Cl3O |
| Molecular Weight (g/mol) | 209.45 |
| MDL Number | MFCD00035937 |
| InChI Key | YBONBWJSFMTXLE-UHFFFAOYSA-N |
| Synonym | benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc |
| PubChem CID | 17944 |
| IUPAC Name | 2,3-dichlorobenzoyl chloride |
| SMILES | ClC(=O)C1=CC=CC(Cl)=C1Cl |
Chemical Identifiers
| 2905-60-4 | |
| 209.45 | |
| YBONBWJSFMTXLE-UHFFFAOYSA-N | |
| 17944 | |
| ClC(=O)C1=CC=CC(Cl)=C1Cl |
| C7H3Cl3O | |
| MFCD00035937 | |
| benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc | |
| 2,3-dichlorobenzoyl chloride |
Specifications
| 2905-60-4 | |
| 140°C | |
| MFCD00035937 | |
| 3261 | |
| YBONBWJSFMTXLE-UHFFFAOYSA-N | |
| 2,3-dichlorobenzoyl chloride | |
| 17944 | |
| ≥98.0% (GC,T) | |
| 2,3-Dichlorobenzoyl Chloride |
| White | |
| C7H3Cl3O | |
| 25 g | |
| benzoyl chloride, dichloro, dichlorobenzoyl chloride, benzoyl chloride, 2,3-dichloro, unii-i50f21ps1n, 2,3-dichlorobenzoylchloride, acmc-1cfin, 2,3-dichlorobenzoyl-chloride, ksc202s8t, 2, 3-dichlorobenzoyl chloride, 2,3-dcoc | |
| ClC(=O)C1=CC=CC(Cl)=C1Cl | |
| 209.45 | |
| 209.45 | |
| Crystals |
Safety and Handling
EINECSNumber : (3)-0062&(3)-1409
TSCA : Yes