missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Dichlorobenzyl Cyanide 98.0+%, TCI America™
$63.38 - $236.73
Chemical Identifiers
| CAS | 3218-45-9 |
|---|---|
| Molecular Formula | C8H5Cl2N |
| Molecular Weight (g/mol) | 186.04 |
| MDL Number | MFCD01861392 |
| InChI Key | CZLINJDTCHACEH-UHFFFAOYSA-N |
| Synonym | 2,3-Dichlorophenylacetonitrile |
| PubChem CID | 229020 |
| IUPAC Name | 2-(2,3-dichlorophenyl)acetonitrile |
| SMILES | ClC1=CC=CC(CC#N)=C1Cl |
Chemical Identifiers
| 3218-45-9 | |
| 186.04 | |
| CZLINJDTCHACEH-UHFFFAOYSA-N | |
| 229020 | |
| ClC1=CC=CC(CC#N)=C1Cl |
| C8H5Cl2N | |
| MFCD01861392 | |
| 2,3-Dichlorophenylacetonitrile | |
| 2-(2,3-dichlorophenyl)acetonitrile |
Specifications
| 3218-45-9 | |
| White-Yellow | |
| C8H5Cl2N | |
| 5 g | |
| 2,3-Dichlorophenylacetonitrile | |
| ClC1=CC=CC(CC#N)=C1Cl | |
| 186.04 | |
| 186.04 | |
| Crystalline Powder |
| 70°C | |
| 115°C | |
| MFCD01861392 | |
| 3439 | |
| CZLINJDTCHACEH-UHFFFAOYSA-N | |
| 2-(2,3-dichlorophenyl)acetonitrile | |
| 229020 | |
| ≥98.0% (GC) | |
| 2,3-Dichlorobenzyl Cyanide |
Safety and Handling
TSCA : No