missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 367710050

 View more versions of this product

Catalog No. AC367710050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 3-Dichlorophenylboronic acid
96.0
242°C to 244°C
Authentic
Glass bottle
MFCD01075703
2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
OB(O)C1=C(Cl)C(Cl)=CC=C1
190.81
190.82
Powder
151169-74-3
100.0
White
97%
C6H5BCl2O2
5 g
TYIKXPOMOYDGCS-UHFFFAOYSA-N
(2,3-dichlorophenyl)boronic acid
2734661
97%

Chemical Identifiers

151169-74-3
190.81
TYIKXPOMOYDGCS-UHFFFAOYSA-N
2734661
OB(O)C1=C(Cl)C(Cl)=CC=C1
C6H5BCl2O2
MFCD01075703
2,3-dichlorophenyl boronic acid, 2,3-dichlorobenzeneboronic acid, 2,3-dichlorophenyl boranediol, boronic acid, 2,3-dichlorophenyl, contains varying amounts of anhydride, pubchem1810, dichlorophenylboronic acid, acmc-1c0ur, dichlorobenzene boronic acid
(2,3-dichlorophenyl)boronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

Recommended Storage : Refrigerator +4°C

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only