missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',3'-Didehydro-3'-deoxythymidine 98.0+%, TCI America™
$98.79 - $326.21
Chemical Identifiers
| CAS | 3056-17-5 |
|---|---|
| Molecular Formula | C10H12N2O4 |
| Molecular Weight (g/mol) | 224.22 |
| MDL Number | MFCD00132921 |
| InChI Key | XNKLLVCARDGLGL-HGXVMFPFNA-N |
| Synonym | stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine |
| PubChem CID | 18283 |
| ChEBI | CHEBI:63581 |
| IUPAC Name | 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| SMILES | CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O |
Chemical Identifiers
| 3056-17-5 | |
| 224.22 | |
| XNKLLVCARDGLGL-HGXVMFPFNA-N | |
| 18283 | |
| 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione |
| C10H12N2O4 | |
| MFCD00132921 | |
| stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine | |
| CHEBI:63581 | |
| CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O |
Specifications
| 3056-17-5 | |
| White | |
| MFCD00132921 | |
| stavudine, sanilvudine, zerit, 2',3'-didehydro-3'-deoxythymidine, estavudina, stavudinum, zerit xr, zerut xr, stavudinum inn-latin, 2',3'-anhydrothymidine | |
| CC1=CN([C@@H]2O[C@H](CO)C=C2)C(=O)NC1=O | |
| 224.22 | |
| CHEBI:63581 | |
| ≥98.0% (HPLC,T) | |
| 2′,3′-Didehydro-3′-deoxythymidine |
| 166°C | |
| C10H12N2O4 | |
| 1 g | |
| XNKLLVCARDGLGL-HGXVMFPFNA-N | |
| 1-[(2R,5S)-5-(hydroxymethyl)-2,5-dihydrofuran-2-yl]-5-methyl-1,2,3,4-tetrahydropyrimidine-2,4-dione | |
| 18283 | |
| 224.22 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : XP2075000
TSCA : No