Learn More
2,3-Difluorotoluene, 97%, Thermo Scientific™
$175.32 - $175.32
Chemical Identifiers
CAS | 3828-49-7 |
---|---|
Molecular Formula | C7H6F2 |
Molecular Weight (g/mol) | 128.12 |
MDL Number | MFCD01631506 |
InChI Key | ZNEHIDGAPGVZSA-UHFFFAOYSA-N |
Synonym | 2,3-difluorotoluene, 1,2-difluoro-3-methyl-benzene, benzene, 1,2-difluoro-3-methyl, 2,3-difluoro methyl benzene, pubchem1610, 2,3-difluoro toluene, acmc-209iz4, 2.3-difluoro methyl benzene, ksc493c6t |
PubChem CID | 2774228 |
IUPAC Name | 1,2-difluoro-3-methylbenzene |
SMILES | CC1=CC=CC(F)=C1F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2395806
|
Thermo Scientific Chemicals
B2395806 |
5 g |
Each for $175.32
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
3828-49-7 | |
128.12 | |
ZNEHIDGAPGVZSA-UHFFFAOYSA-N | |
2774228 | |
CC1=CC=CC(F)=C1F |
C7H6F2 | |
MFCD01631506 | |
2,3-difluorotoluene, 1,2-difluoro-3-methyl-benzene, benzene, 1,2-difluoro-3-methyl, 2,3-difluoro methyl benzene, pubchem1610, 2,3-difluoro toluene, acmc-209iz4, 2.3-difluoro methyl benzene, ksc493c6t | |
1,2-difluoro-3-methylbenzene |
Specifications
3828-49-7 | |
115°C to 116°C | |
Aromatic | |
1.4522 | |
5 g | |
3235341 | |
ZNEHIDGAPGVZSA-UHFFFAOYSA-N | |
1,2-difluoro-3-methylbenzene | |
2774228 | |
97% |
1.12 | |
15°C (59°F) | |
C7H6F2 | |
MFCD01631506 | |
UN2388 | |
2,3-difluorotoluene, 1,2-difluoro-3-methyl-benzene, benzene, 1,2-difluoro-3-methyl, 2,3-difluoro methyl benzene, pubchem1610, 2,3-difluoro toluene, acmc-209iz4, 2.3-difluoro methyl benzene, ksc493c6t | |
CC1=CC=CC(F)=C1F | |
128.12 | |
128.12 | |
2,3-Difluorotoluene |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
H225-H302-H315-H318-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLUOROTOLUENES
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures