missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3-Dihydro-1-benzofuran-7-sulfonyl chloride, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 953408-82-7 |
---|---|
Molecular Formula | C8H7ClO3S |
Molecular Weight (g/mol) | 218.65 |
MDL Number | MFCD11109309 |
InChI Key | UNECAYDZDLGSLP-UHFFFAOYSA-N |
Synonym | 2,3-dihydrobenzofuran-7-sulfonyl chloride, 2,3-dihydro-benzofuran-7-sulfonyl chloride, 7-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzo b furan-7-sulfonyl chloride, chloro-2,3-dihydrobenzo 2,3-b furan-7-ylsulfone |
PubChem CID | 18616199 |
IUPAC Name | 2,3-dihydro-1-benzofuran-7-sulfonyl chloride |
SMILES | ClS(=O)(=O)C1=CC=CC2=C1OCC2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
CC00903CB
|
Thermo Scientific Chemicals
CC00903CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC00903DA
|
Thermo Scientific Chemicals
CC00903DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
CC00903DE
|
Thermo Scientific Chemicals
CC00903DE |
5g | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
953408-82-7 | |
218.65 | |
UNECAYDZDLGSLP-UHFFFAOYSA-N | |
18616199 | |
ClS(=O)(=O)C1=CC=CC2=C1OCC2 |
C8H7ClO3S | |
MFCD11109309 | |
2,3-dihydrobenzofuran-7-sulfonyl chloride, 2,3-dihydro-benzofuran-7-sulfonyl chloride, 7-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzo b furan-7-sulfonyl chloride, chloro-2,3-dihydrobenzo 2,3-b furan-7-ylsulfone | |
2,3-dihydro-1-benzofuran-7-sulfonyl chloride |
Specifications
953408-82-7 | |
Amber Glass Bottle | |
MFCD11109309 | |
2,3-dihydrobenzofuran-7-sulfonyl chloride, 2,3-dihydro-benzofuran-7-sulfonyl chloride, 7-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzo b furan-7-sulfonyl chloride, chloro-2,3-dihydrobenzo 2,3-b furan-7-ylsulfone | |
ClS(=O)(=O)C1=CC=CC2=C1OCC2 | |
218.65 | |
218.66 | |
2, 3-Dihydro-1-benzofuran-7-sulfonyl chloride |
97% | |
C8H7ClO3S | |
250mg | |
UNECAYDZDLGSLP-UHFFFAOYSA-N | |
2,3-dihydro-1-benzofuran-7-sulfonyl chloride | |
18616199 | |
97% |
Safety and Handling
GHS Signal Word: Danger