missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxy-5-methyl-1,4-benzoquinone, 98%

CAS: 605-94-7 | C9H10O4 | 182.18 g/mol

$99.15 - $99.15

Chemical Identifiers

CAS 605-94-7
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00001595
InChI Key UIXPTCZPFCVOQF-UHFFFAOYSA-N
Synonym 2,3-dimethoxy-5-methyl-1,4-benzoquinone, coenzyme q0, ubiquinone 0, 2,3-dimethoxy-5-methyl-p-benzoquinone, ubiquinone q0, ubiquinone-o, ubiquinone-0, coq0, 2-methyl-4,5-dimethoxy-p-quinone, 2,3-dimethoxy-5-methylbenzoquinone
PubChem CID 69068
ChEBI CHEBI:27906
IUPAC Name 2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
SMILES COC1=C(OC)C(=O)C(C)=CC1=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442200010
SDS View Documents
Thermo Scientific Chemicals
442200010
1 g
Each for $99.15
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

605-94-7
182.18
UIXPTCZPFCVOQF-UHFFFAOYSA-N
69068
2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
C9H10O4
MFCD00001595
2,3-dimethoxy-5-methyl-1,4-benzoquinone, coenzyme q0, ubiquinone 0, 2,3-dimethoxy-5-methyl-p-benzoquinone, ubiquinone q0, ubiquinone-o, ubiquinone-0, coq0, 2-methyl-4,5-dimethoxy-p-quinone, 2,3-dimethoxy-5-methylbenzoquinone
CHEBI:27906
COC1=C(OC)C(=O)C(C)=CC1=O

Specifications

605-94-7
100.0
Red to Orange
97.5% min. (HPLC)
C9H10O4
1 g
UIXPTCZPFCVOQF-UHFFFAOYSA-N
2,3-dimethoxy-5-methylcyclohexa-2,5-diene-1,4-dione
69068
182.18
Crystalline Powder or Needles
97.5
59°C to 61°C
Authentic
Glass Bottle
MFCD00001595
2,3-dimethoxy-5-methyl-1,4-benzoquinone, coenzyme q0, ubiquinone 0, 2,3-dimethoxy-5-methyl-p-benzoquinone, ubiquinone q0, ubiquinone-o, ubiquinone-0, coq0, 2-methyl-4,5-dimethoxy-p-quinone, 2,3-dimethoxy-5-methylbenzoquinone
COC1=C(OC)C(=O)C(C)=CC1=O
182.18
CHEBI:27906
98%
2, 3-Dimethoxy-5-methyl-1, 4-benzoquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-100-8

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only