missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dimethoxyphenol, 99%

CAS: 5150-42-5 | C8H10O3 | 154.17 g/mol

$177.35 - $177.35

Chemical Identifiers

CAS 5150-42-5
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00008366
InChI Key QSZCGGBDNYTQHH-UHFFFAOYSA-N
Synonym dimethoxyphenol, phenol, dimethoxy, unii-ow147i6c84, 1-hydroxy-2,3-dimethoxybenzene, pyrogallol 1,2-dimethyl ether, phenol, 2,3-dimethoxy, phenol,3-dimethoxy, 2,3-di-methoxyphenol, 2,3-dim ethoxyphenol, 2,3-dimethoxy phenol
PubChem CID 78828
IUPAC Name 2,3-dimethoxyphenol
SMILES COC1=CC=CC(=C1OC)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC155250050
SDS View Documents
Thermo Scientific Chemicals
155250050
5 g Glass bottle
Each for $177.35
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5150-42-5
154.17
QSZCGGBDNYTQHH-UHFFFAOYSA-N
78828
COC1=CC=CC(=C1OC)O
C8H10O3
MFCD00008366
dimethoxyphenol, phenol, dimethoxy, unii-ow147i6c84, 1-hydroxy-2,3-dimethoxybenzene, pyrogallol 1,2-dimethyl ether, phenol, 2,3-dimethoxy, phenol,3-dimethoxy, 2,3-di-methoxyphenol, 2,3-dim ethoxyphenol, 2,3-dimethoxy phenol
2,3-dimethoxyphenol

Specifications

5150-42-5
233.0°C to 234.0°C
Authentic
Glass bottle
1.5385 to 1.5405
MFCD00008366
06,1081
QSZCGGBDNYTQHH-UHFFFAOYSA-N
2,3-dimethoxyphenol
78828
99%
2, 3-Dimethoxyphenol, 99%
Orange to Brown
108°C
99%
C8H10O3
(CH3O)2C6H3OH
5 g
dimethoxyphenol, phenol, dimethoxy, unii-ow147i6c84, 1-hydroxy-2,3-dimethoxybenzene, pyrogallol 1,2-dimethyl ether, phenol, 2,3-dimethoxy, phenol,3-dimethoxy, 2,3-di-methoxyphenol, 2,3-dim ethoxyphenol, 2,3-dimethoxy phenol
COC1=CC=CC(=C1OC)O
154.17
154.17
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-922-2

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only