missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Dimethyl-1-phenyl-5-pyrazolone 99.0+%, TCI America™

$39.52 - $269.76

Chemical Identifiers

CAS 60-80-0
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD00003146
InChI Key VEQOALNAAJBPNY-UHFFFAOYSA-N
Synonym antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
PubChem CID 2206
ChEBI CHEBI:31225
IUPAC Name 1,5-dimethyl-2-phenylpyrazol-3-one
SMILES CC1=CC(=O)N(N1C)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D187625G
View Documents
TCI America
D187625G
25 g
Each for $39.52
 
D1876500G
View Documents
TCI America
D1876500G
500 g
Each for $269.76
 
Description

Description

Specifications

Chemical Identifiers

60-80-0
188.23
VEQOALNAAJBPNY-UHFFFAOYSA-N
2206
1,5-dimethyl-2-phenylpyrazol-3-one
C11H12N2O
MFCD00003146
antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
CHEBI:31225
CC1=CC(=O)N(N1C)C2=CC=CC=C2

Specifications

60-80-0
White
C11H12N2O
25 g
VEQOALNAAJBPNY-UHFFFAOYSA-N
1,5-dimethyl-2-phenylpyrazol-3-one
2206
188.23
Crystalline Powder
113°C
319°C
MFCD00003146
antipyrine, phenazone, antipyrin, phenazon, analgesine, anodynine, anodynin, azophen, fenazone, 2,3-dimethyl-1-phenyl-5-pyrazolone
CC1=CC(=O)N(N1C)C2=CC=CC=C2
188.23
CHEBI:31225
≥99.0% (GC,T)
2,3-Dimethyl-1-phenyl-5-pyrazolone
Safety and Handling

Safety and Handling

EINECSNumber : (9)-1252

RTECSNumber : CD2450000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'