Learn More
2-(3-Fluorophenyl)ethylamine, ≥97%, Thermo Scientific Chemicals
$62.37 - $209.10
Chemical Identifiers
CAS | 404-70-6 |
---|---|
Molecular Formula | C8H11FN |
Molecular Weight (g/mol) | 140.18 |
MDL Number | MFCD00075376 |
InChI Key | AUCVZEYHEFAWHO-UHFFFAOYSA-O |
Synonym | 3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine |
PubChem CID | 533928 |
IUPAC Name | 2-(3-fluorophenyl)ethanamine |
SMILES | [NH3+]CCC1=CC=CC(F)=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6476206
|
Thermo Scientific Chemicals
H6476206 |
5 g |
N/A
|
|
|||||
AAH6476214
|
Thermo Scientific Chemicals
H6476214 |
25 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
404-70-6 | |
140.18 | |
AUCVZEYHEFAWHO-UHFFFAOYSA-O | |
533928 | |
[NH3+]CCC1=CC=CC(F)=C1 |
C8H11FN | |
MFCD00075376 | |
3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine | |
2-(3-fluorophenyl)ethanamine |
Specifications
404-70-6 | |
87°C (15 mmHg) | |
C8H11FN | |
MFCD00075376 | |
UN2735 | |
3-fluorophenethylamine, 2-3-fluorophenyl ethylamine, 2-3-fluoro-phenyl-ethylamine, 2-3-fluorophenyl ethanamine, 2-3-fluorophenyl ethan-1-amine, benzeneethanamine, 3-fluoro, m-fluorophenethylamine, 3-flourophenethylamine, m-fluorophenylethylamine, 3-fluoro phenethylamine | |
[NH3+]CCC1=CC=CC(F)=C1 | |
140.18 | |
139.17 | |
Liquid |
1.066 g/mL | |
83°C (181°F) | |
1.509 | |
5 g | |
3045789 | |
AUCVZEYHEFAWHO-UHFFFAOYSA-O | |
2-(3-fluorophenyl)ethanamine | |
533928 | |
≥97% | |
2-(3-Fluorophenyl)ethylamine |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only