missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3-Pentanedione, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 221475000

Catalog No. AC221475000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2, 3-Pentanedione
600-14-6
100.0
110.0°C to 112.0°C
Authentic
Glass bottle
1.4020 to 1.4060
MFCD00009313
01,776
2,3-pentanedione, acetylpropionyl, acetyl propionyl, 2,3-pentadione, acetylpropionyl van, unii-k4wbe45scm, acetyl propionyl natural, 2,3-pentane-dione, pentan-2,3-dione, ethyl methyl diketone
TZMFJUDUGYTVRY-UHFFFAOYSA-N
pentane-2,3-dione
11747
100.12
97%
96.0
0.9580g/mL
18°C
96% min. (GC)
C5H8O2
C2H5COCOCH3
500mL
0.958
Solubility in water: 60g/L (15°C). Other solubilities: miscible with ethanol, glycerol and, propylene glycol, ether
CCC(=O)C(C)=O
100.12
CHEBI:52774
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 209-984-8

RTECSNumber : SA1850000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only