Learn More
2(3)-tert-Butyl-4-methoxyphenol, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2353030
Description
2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2(3)-tert-Butyl-4-methoxyphenol is used as an industrial antioxidant. It is used as a food additive, preservative in food packaging, animal feed, cosmetics, rubber, and petroleum products.
Solubility
Soluble in ethanol, methanol, chloroform, fats, and oils. Insoluble in water.
Notes
Store in the dark place. Store away from light, oxidizing agents.
Specifications
2(3)-tert-Butyl-4-methoxyphenol | |
55°C to 65°C | |
MFCD01779059 | |
Light sensitive | |
3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
2-tert-butyl-4-methoxyphenol | |
8456 | |
180.25 |
25013-16-5 | |
C11H16O2 | |
250 g | |
14,1547 | |
Soluble in ethanol,methanol,chloroform,fats,and oils. Insoluble in water. | |
CC(C)(C)C1=C(C=CC(=C1)OC)O | |
180.247 | |
CHEBI:76358 | |
96% |
Chemical Identifiers
25013-16-5 | |
180.247 | |
MRBKEAMVRSLQPH-UHFFFAOYSA-N | |
8456 | |
2-tert-butyl-4-methoxyphenol |
C11H16O2 | |
MFCD01779059 | |
3-tert-butyl-4-hydroxyanisole, 4-hydroxy-3-tert-butylanisole, 2-tert-butyl-4-methoxyphenol, 3-bha, 4-methoxy-2-tert-butylphenol, 3-t-butyl-4-hydroxyanisole, phenol, 2-1,1-dimethylethyl-4-methoxy, 2-1,1-dimethylethyl-4-methoxyphenol, 2-butyl-4-hydroxyanisole, phenol, 2-tert-butyl-4-methoxy | |
CHEBI:76358 | |
CC(C)(C)C1=C(C=CC(=C1)OC)O |
Safety and Handling
GHS H Statement
H351-H302
Suspected of causing cancer.
Harmful if swallowed.
P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302-H315-H319-H335-H350
EINECSNumber : 246-563-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only