missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[3-(Trifluoromethyl)phenyl]ethylamine, 98%, Thermo Scientific Chemicals

$60.05 - $815.96

Chemical Identifiers

CAS 52516-30-0
Molecular Formula C9H10F3N
Molecular Weight (g/mol) 189.181
MDL Number MFCD00040756
InChI Key BPVYCXMGJPKOTQ-UHFFFAOYSA-N
Synonym 2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
PubChem CID 104223
IUPAC Name 2-[3-(trifluoromethyl)phenyl]ethanamine
SMILES C1=CC(=CC(=C1)C(F)(F)F)CCN
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6408403
View Documents
Thermo Scientific Chemicals
H6408403
1 g
Each for $60.05
 
AAH6408406
View Documents
Thermo Scientific Chemicals
H6408406
5 g
Each for $208.01
 
AAH6408414
View Documents
Thermo Scientific Chemicals
H6408414
25 g
Each for $815.96
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

52516-30-0
189.181
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
104223
C1=CC(=CC(=C1)C(F)(F)F)CCN
C9H10F3N
MFCD00040756
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
2-[3-(trifluoromethyl)phenyl]ethanamine

Specifications

52516-30-0
1.18 g/mL
C9H10F3N
MFCD00040756
UN2735
Light sensitive
BPVYCXMGJPKOTQ-UHFFFAOYSA-N
2-[3-(trifluoromethyl)phenyl]ethanamine
104223
98%
2-[3-(Trifluoromethyl)phenyl]ethylamine
Colorless to Yellow
113°C (22 mmHg)
1.4645
1 g
4398117
2-3-trifluoromethyl phenyl ethanamine, 3-trifluoromethyl phenethylamine, 2-3-trifluoromethylphenyl ethylamine, 2-3-trifluoromethyl phenyl ethylamine, phenethylamine, m-trifluoromethyl, m-trifluoromethylphenethylamine, m-trifluoromethyl phenethylamine, benzeneethanamine, 3-trifluoromethyl, 2-3-trifluoromethylphenyl-ethylamine
C1=CC(=CC(=C1)C(F)(F)F)CCN
189.181
189.18
Liquid
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 257-988-3

RTECSNumber : SI2525000

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only