missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,4-Trimethyl-2-pentene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 162630050

 View more versions of this product

Catalog No. AC162630050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4, 4-Trimethyl-2-pentene
107-40-4
100.0
Colorless to Yellow
104.0°C
Authentic
Glass bottle
1.4150 to 1.4170
MFCD00008902
01, III, 848
2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208
CC(=CC(C)(C)C)C
112.21
112.21
Liquid
98%
97.5
-106.0°C
0.7200g/mL
2°C
98%
C8H16
(CH3)3CCH=C(CH3)2
5 mL
0.72
LAAVYEUJEMRIGF-UHFFFAOYSA-N
2,4,4-trimethylpent-2-ene
7869
98%

Chemical Identifiers

107-40-4
112.21
LAAVYEUJEMRIGF-UHFFFAOYSA-N
7869
CC(=CC(C)(C)C)C
C8H16
MFCD00008902
2,4,4-trimethyl-2-pentene, 2-pentene, 2,4,4-trimethyl, 2,2,4-trimethyl-3-pentene, 2,4,4-trimethylpentene-2, unii-548r0du21i, beta-diisobutylene, propene, 1-tert-butyl-2-methyl, acmc-1cujo, dsstox_cid_6766, dsstox_rid_78208
2,4,4-trimethylpent-2-ene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with lon

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or ha

GHS Signal Word: Danger

EINECSNumber : 203-488-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only