missing translation for 'onlineSavingsMsg'
Learn More

2,4,5-Triamino-6-hydroxypyrimidine sulfate, 94%

CAS: 35011-47-3 | C4H9N5O5S | 239.206 g/mol

$106.27 - $324.82

Chemical Identifiers

CAS 35011-47-3
Molecular Formula C4H9N5O5S
Molecular Weight (g/mol) 239.206
MDL Number MFCD00012785
InChI Key RSKNEEODWFLVFF-UHFFFAOYSA-N
Synonym 2,4,5-triamino-6-hydroxypyrimidine sulfate, 6-hydroxy-2,4,5-triaminopyrimidine sulfate, 2,5,6-triaminopyrimidin-4 3h-one sulfate, unii-9sz7zx1h0l, sulfuric acid; triaminopyrimidin-4-ol, 2,5,6-triamino-4-pyrimidinol sulfate, 4 1h-pyrimidinone, 2,5,6-triamino-, sulfate, 2,5,6-triaminopyrimidin-4 1h-one sulphate, 2,5,6-triaminopyrimidin-4-ol sulfate, 9sz7zx1h0l
PubChem CID 296856
IUPAC Name sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one
SMILES C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2198914
View Documents
Thermo Scientific Chemicals
B2198914
25 g N/A
Only null left
 
AAB2198922
View Documents
Thermo Scientific Chemicals
B2198922
100 g N/A
Only null left
 
Description

Description

2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Triamino-6-hydroxypyrimidine sulfate used as an intermediate for synthetic organic chemistry.

Solubility
Soluble in DMSO.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

35011-47-3
239.206
RSKNEEODWFLVFF-UHFFFAOYSA-N
296856
C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O
C4H9N5O5S
MFCD00012785
2,4,5-triamino-6-hydroxypyrimidine sulfate, 6-hydroxy-2,4,5-triaminopyrimidine sulfate, 2,5,6-triaminopyrimidin-4 3h-one sulfate, unii-9sz7zx1h0l, sulfuric acid; triaminopyrimidin-4-ol, 2,5,6-triamino-4-pyrimidinol sulfate, 4 1h-pyrimidinone, 2,5,6-triamino-, sulfate, 2,5,6-triaminopyrimidin-4 1h-one sulphate, 2,5,6-triaminopyrimidin-4-ol sulfate, 9sz7zx1h0l
sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one

Specifications

35011-47-3
C4H9N5O5S
25 g
2,4,5-triamino-6-hydroxypyrimidine sulfate, 6-hydroxy-2,4,5-triaminopyrimidine sulfate, 2,5,6-triaminopyrimidin-4 3h-one sulfate, unii-9sz7zx1h0l, sulfuric acid; triaminopyrimidin-4-ol, 2,5,6-triamino-4-pyrimidinol sulfate, 4 1h-pyrimidinone, 2,5,6-triamino-, sulfate, 2,5,6-triaminopyrimidin-4 1h-one sulphate, 2,5,6-triaminopyrimidin-4-ol sulfate, 9sz7zx1h0l
RSKNEEODWFLVFF-UHFFFAOYSA-N
sulfuric acid;2,5,6-triamino-1H-pyrimidin-4-one
296856
97%
>300°C
MFCD00012785
4621048
Soluble in DMSO.
C1(=C(NC(=NC1=O)N)N)N.OS(=O)(=O)O
239.206
239.21
2,4,5-Triamino-6-hydroxypyrimidine sulfate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UW7175000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only