Learn More
2,4,5-Trichloropyrimidine, 98%, Thermo Scientific Chemicals
$269.60 - $269.60
Chemical Identifiers
CAS | 5750-76-5 |
---|---|
Molecular Formula | C4HCl3N2 |
Molecular Weight (g/mol) | 183.416 |
MDL Number | MFCD03788200 |
InChI Key | GIKMWFAAEIACRF-UHFFFAOYSA-N |
Synonym | 2,5,6-trichloropyrimidine, 2,4,5-trichloro-pyrimidine, trichloropyrimidine, pyrimidine, 2,4,5-trichloro, 2,4,5-trichlorpyrimidin, pubchem5302, zlchem 1349, pyrimidine, trichloro, acmc-209lys, 2,5-trichloropyrimidine |
PubChem CID | 237259 |
IUPAC Name | 2,4,5-trichloropyrimidine |
SMILES | C1=C(C(=NC(=N1)Cl)Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1950606
|
Thermo Scientific Chemicals
L1950606 |
5 g |
N/A
|
|
|||||
Description
2,4,5-Trichloropyrimidine is used in the synthesis of potent and selective anaplastic lymphoma kinase (ALK-5) inhibitors, used as an anti-tumor treatment. It is also used in the synthesis of EGFR inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,5-Trichloropyrimidine is used in the synthesis of potent and selective anaplastic lymphoma kinase (ALK-5) inhibitors, used as an anti-tumor treatment. It is also used in the synthesis of EGFR inhibitors.
Solubility
Not miscible or difficult to mix in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
5750-76-5 | |
183.416 | |
GIKMWFAAEIACRF-UHFFFAOYSA-N | |
237259 | |
C1=C(C(=NC(=N1)Cl)Cl)Cl |
C4HCl3N2 | |
MFCD03788200 | |
2,5,6-trichloropyrimidine, 2,4,5-trichloro-pyrimidine, trichloropyrimidine, pyrimidine, 2,4,5-trichloro, 2,4,5-trichlorpyrimidin, pubchem5302, zlchem 1349, pyrimidine, trichloro, acmc-209lys, 2,5-trichloropyrimidine | |
2,4,5-trichloropyrimidine |
Specifications
5750-76-5 | |
84°C (1 mmHg) | |
C4HCl3N2 | |
MFCD03788200 | |
UN1760 | |
2,5,6-trichloropyrimidine, 2,4,5-trichloro-pyrimidine, trichloropyrimidine, pyrimidine, 2,4,5-trichloro, 2,4,5-trichlorpyrimidin, pubchem5302, zlchem 1349, pyrimidine, trichloro, acmc-209lys, 2,5-trichloropyrimidine | |
GIKMWFAAEIACRF-UHFFFAOYSA-N | |
2,4,5-trichloropyrimidine | |
237259 | |
98% |
1.6 | |
>110°C (230°F) | |
1.573 | |
5 g | |
4449 | |
Not miscible or difficult to mix in water. | |
C1=C(C(=NC(=N1)Cl)Cl)Cl | |
183.416 | |
183.43 | |
2,4,5-Trichloropyrimidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber : 000-000-0
TSCA : No
RUO – Research Use Only