missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trifluoroaniline, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1676506

 View more versions of this product

Catalog No. AAA1676506


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4,5-Trifluoroaniline
59°C to 63°C
MFCD00007649
UN2811
2,4,5-trifluorophenylamine, benzenamine, 2,4,5-trifluoro, pubchem2469, 2,4,5-triflouroaniline, 2,4,5-trifloro aniline, acmc-1cs4m, 2,4,5-trifluoro-aniline, 2,4,5-trifluoro aniline, ksc497k9p, 2,4,5-trifluoro-phenylamine
C1=C(C(=CC(=C1F)F)F)N
147.1
147.1
367-34-0
C6H4F3N
5 g
2085734
QMYVWJVVVMIBMM-UHFFFAOYSA-N
2,4,5-trifluoroaniline
94953
98%

Chemical Identifiers

367-34-0
147.1
QMYVWJVVVMIBMM-UHFFFAOYSA-N
94953
C1=C(C(=CC(=C1F)F)F)N
C6H4F3N
MFCD00007649
2,4,5-trifluorophenylamine, benzenamine, 2,4,5-trifluoro, pubchem2469, 2,4,5-triflouroaniline, 2,4,5-trifloro aniline, acmc-1cs4m, 2,4,5-trifluoro-aniline, 2,4,5-trifluoro aniline, ksc497k9p, 2,4,5-trifluoro-phenylamine
2,4,5-trifluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H318

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 206-692-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only