missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

2,4,5-Trihydroxybenzaldehyde, 97%, C7H6O4, CAS Number-35094-87-2, benzaldehyde, 2,4,5-trihydroxy, benzaldehyde,2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxybenzaldehyde, 2,4,5-trihydroxy-benzaldehyde, 5g, 2250084, 154.12, H315-H319-H335

$291.60 - $967.04

Chemical Identifiers

CAS 35094-87-2
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.12
MDL Number MFCD00016592
InChI Key WNCNWLVQSHZVKV-UHFFFAOYSA-N
PubChem CID 643387
IUPAC Name 2,4,5-trihydroxybenzaldehyde
SMILES OC1=CC(O)=C(C=O)C=C1O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1328303
View Documents
Thermo Scientific Chemicals
A1328303
1 g N/A
 
AAA1328306
View Documents
Thermo Scientific Chemicals
A1328306
5 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35094-87-2
154.12
WNCNWLVQSHZVKV-UHFFFAOYSA-N
2,4,5-trihydroxybenzaldehyde
C7H6O4
MFCD00016592
643387
OC1=CC(O)=C(C=O)C=C1O

Specifications

35094-87-2
C7H6O4
1 g
Air Sensitive
WNCNWLVQSHZVKV-UHFFFAOYSA-N
2,4,5-trihydroxybenzaldehyde
643387
97%
∼230°C (decomposition)
MFCD00016592
2250084
benzaldehyde, 2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxy-benzaldehyde, benzaldehyde,2,4,5-trihydroxy, 2,4,5-trihydroxybenzaldehyde
OC1=CC(O)=C(C=O)C=C1O
154.12
154.12
2,4,5-Trihydroxybenzaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only