missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,5-Trihydroxybenzaldehyde, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 345940010

 View more versions of this product

Catalog No. AC345940010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4, 5-Trihydroxybenzaldehyde
98.5
226°C to 227°C
Authentic
Glass bottle
MFCD00016592
08, IV, 2715
Solubility in water: soluble. Other solubilities: soluble in diethyl ether
OC1=CC(O)=C(C=O)C=C1O
154.12
154.12
Crystalline Powder
35094-87-2
100.0
Brown to Yellow or Khaki
98.5% min. (HPLC)
C7H6O4
1 g
benzaldehyde, 2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxy-benzaldehyde, benzaldehyde,2,4,5-trihydroxy, 2,4,5-trihydroxybenzaldehyde
WNCNWLVQSHZVKV-UHFFFAOYSA-N
2,4,5-trihydroxybenzaldehyde
643387
99%

Chemical Identifiers

35094-87-2
154.12
WNCNWLVQSHZVKV-UHFFFAOYSA-N
2,4,5-trihydroxybenzaldehyde
C7H6O4
MFCD00016592
643387
OC1=CC(O)=C(C=O)C=C1O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only