missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,5-Trihydroxybenzaldehyde 98.0+%, TCI America™
Supplier: TCI America T17661G
Specifications
2,4,5-Trihydroxybenzaldehyde | |
Green-Yellow | |
MFCD00016592 | |
benzaldehyde, 2,4,5-trihydroxy, acmc-1ag53, 4,5-dihydroxy-salicylaldehyde, 2,4,5-trihydroxy-benzaldehyde, benzaldehyde,2,4,5-trihydroxy, 2,4,5-trihydroxybenzaldehyde | |
OC1=CC(O)=C(C=O)C=C1O | |
154.12 | |
154.12 | |
Crystalline Powder |
35094-87-2 | |
C7H6O4 | |
1 g | |
WNCNWLVQSHZVKV-UHFFFAOYSA-N | |
2,4,5-trihydroxybenzaldehyde | |
643387 | |
≥98.0% (GC) |
Chemical Identifiers
35094-87-2 | |
154.12 | |
WNCNWLVQSHZVKV-UHFFFAOYSA-N | |
2,4,5-trihydroxybenzaldehyde |
C7H6O4 | |
MFCD00016592 | |
643387 | |
OC1=CC(O)=C(C=O)C=C1O |
Safety and Handling
TSCA : No