missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4,5-Trimethoxybenzoic Acid 98.0+%, TCI America™
$43.44 - $104.68
Chemical Identifiers
CAS | 490-64-2 |
---|---|
Molecular Formula | C10H11O5 |
Molecular Weight (g/mol) | 211.19 |
MDL Number | MFCD00002435 |
InChI Key | KVZUCOGWKYOPID-UHFFFAOYSA-M |
Synonym | asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid |
PubChem CID | 10276 |
IUPAC Name | 2,4,5-trimethoxybenzoate |
SMILES | COC1=CC(OC)=C(C=C1OC)C([O-])=O |
Chemical Identifiers
490-64-2 | |
211.19 | |
KVZUCOGWKYOPID-UHFFFAOYSA-M | |
10276 | |
COC1=CC(OC)=C(C=C1OC)C([O-])=O |
C10H11O5 | |
MFCD00002435 | |
asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid | |
2,4,5-trimethoxybenzoate |
Specifications
490-64-2 | |
White-Yellow | |
C10H11O5 | |
5 g | |
KVZUCOGWKYOPID-UHFFFAOYSA-M | |
2,4,5-trimethoxybenzoate | |
10276 | |
≥98.0% (GC,T) | |
2,4,5-Trimethoxybenzoic Acid |
144°C | |
300°C | |
MFCD00002435 | |
asaronic acid, benzoic acid, 2,4,5-trimethoxy, asarylic acid, calamonic acid, acmc-209ket, 2,4,5-trimethoxybenzoicacid, ksc489o7d, 2,4,5-trimethoxy-benzoic acid, 2,4,5-trimethoxybenzoic acid, alpha-hydroxyphenylacetic acid; amygdalic acid; alpha-hydroxyphenylacetic acid; alpha-hydroxy-benzeneacetic acid | |
COC1=CC(OC)=C(C=C1OC)C([O-])=O | |
211.19 | |
212.20 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-4610
TSCA : Yes