missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,5-Trimethylbenzoic Acid 97.0+%, TCI America™
$233.00 - $924.57
Chemical Identifiers
| CAS | 528-90-5 |
|---|---|
| Molecular Formula | C10H12O2 |
| Molecular Weight (g/mol) | 164.20 |
| MDL Number | MFCD00075863 |
| InChI Key | QENJZWZWAWWESF-UHFFFAOYSA-N |
| PubChem CID | 10714 |
| IUPAC Name | 2,4,5-trimethylbenzoic acid |
| SMILES | CC1=CC(C)=C(C=C1C)C(O)=O |
Chemical Identifiers
| 528-90-5 | |
| 164.20 | |
| QENJZWZWAWWESF-UHFFFAOYSA-N | |
| 2,4,5-trimethylbenzoic acid |
| C10H12O2 | |
| MFCD00075863 | |
| 10714 | |
| CC1=CC(C)=C(C=C1C)C(O)=O |
Specifications
| 528-90-5 | |
| White-Yellow | |
| MFCD00075863 | |
| QENJZWZWAWWESF-UHFFFAOYSA-N | |
| 2,4,5-trimethylbenzoic acid | |
| 10714 | |
| ≥97.0% (GC,T) | |
| 2,4,5-Trimethylbenzoic Acid |
| 151°C | |
| C10H12O2 | |
| 5 g | |
| CC1=CC(C)=C(C=C1C)C(O)=O | |
| 164.20 | |
| 164.20 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : DI0887000
TSCA : No