missing translation for 'onlineSavingsMsg'
Learn More

2,4,6,8-Tetramethylcyclotetrasiloxane 95.0+%, TCI America™

Supplier:  TCI America T2076100G

 View more versions of this product

Catalog No. T2076100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4,6,8-Tetramethylcyclotetrasiloxane
Colorless
C4H12O4Si4
100 g
2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane
C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C
236.476
240.51
Liquid
2370-88-9
134°C
MFCD00039567
1993
WZJUBBHODHNQPW-UHFFFAOYSA-N
2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene
6327421
≥95.0% (GC)

Chemical Identifiers

2370-88-9
236.476
WZJUBBHODHNQPW-UHFFFAOYSA-N
6327421
C[Si-]1O[Si-]([O+]=[Si]([O+]=[Si](O1)C)C)C
C4H12O4Si4
MFCD00039567
2,4,6,8-tetramethylcyclotetrasiloxane, 1,3,5,7-tetramethylcyclotetrasiloxane, tetramethylcyclotetrasiloxane, tmcts, cyclotetrasiloxane, 2,4,6,8-tetramethyl, 2,4,6,8-tetramethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, 1,2,5,7-tetramethylcyclotetrasiloxane, 2,4,6,8-tetramethyl-cyclotetrasiloxan, 2,4,6,8-tetramethylcyclotetrasiloxne, 1,3,5,7-tetramethyl cyclotetrasiloxane
2,4,6,8-tetramethyl-5,7-dioxa-1,3-dioxonia-2,4-disila-6,8-disilanidacycloocta-1,3-diene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (7)-0477

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions