missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trichlorophenol, 98%, Thermo Scientific Chemicals

$100.14 - $203.70

Chemical Identifiers

CAS 88-06-2
Molecular Formula C6H3Cl3O
Molecular Weight (g/mol) 197.45
MDL Number MFCD00002172
InChI Key LINPIYWFGCPVIE-UHFFFAOYSA-N
Synonym dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz
PubChem CID 6914
ChEBI CHEBI:28755
IUPAC Name 2,4,6-trichlorophenol
SMILES C1=C(C=C(C(=C1Cl)O)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC139481000
SDS View Documents
Thermo Scientific Chemicals
139481000
100 g Glass bottle
Each for $100.14
 
AC139485000
SDS View Documents
Thermo Scientific Chemicals
139485000
500 g Glass bottle
Each for $203.70
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

88-06-2
197.45
LINPIYWFGCPVIE-UHFFFAOYSA-N
6914
2,4,6-trichlorophenol
C6H3Cl3O
MFCD00002172
dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz
CHEBI:28755
C1=C(C=C(C(=C1Cl)O)Cl)Cl

Specifications

88-06-2
100.0
Beige to Brown
99°C
97.5% min. (GC)
C6H3Cl3O
MFCD00002172
06, 190
dowicide 2s, phenachlor, phenol, 2,4,6-trichloro, omal, phenaclor, rcra waste number u231, dowcide 2s, 1,3,5-trichloro-2-hydroxybenzene, 2,4,6-trichlorfenol, unii-mhs8c5bauz
LINPIYWFGCPVIE-UHFFFAOYSA-N
2,4,6-trichlorophenol
6914
197.45
Crystalline Mass or Crystalline Powder and Chunks
97.5
64.0°C to 68.0°C
246.0°C
Authentic
Glass bottle
Cl3C6H2OH
100 g
15, 9809
Solubility in water: 0.8g/L. Other solubilities: soluble in aromatic solvents and caustic soda sol.,soluble in (g/100g) acetone 525; benzene 113;,ccl4 37; diacetone alcohol 335; ether 354;,methanol 525; pine oil 163; toluene 100;,turpentine 37
C1=C(C=C(C(=C1Cl)O)Cl)Cl
197.45
CHEBI:28755
98%
2, 4, 6-Trichlorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/p

GHS Signal Word: Warning

EINECSNumber : 201-795-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only