missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzoic acid, 98%

CAS: 570-02-5 | C10H12O5 | 212.20 g/mol

$201.25 - $702.59

Chemical Identifiers

CAS 570-02-5
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.20
MDL Number MFCD00016497
InChI Key JATAKEDDMQNPOQ-UHFFFAOYSA-N
Synonym benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid
PubChem CID 68441
IUPAC Name 2,4,6-trimethoxybenzoic acid
SMILES COC1=CC(OC)=C(C(O)=O)C(OC)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0499706
View Documents
Thermo Scientific Chemicals
L0499706
5 g
Each for $201.25
Only null left
 
AAL0499714
View Documents
Thermo Scientific Chemicals
L0499714
25 g
Each for $702.59
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

570-02-5
212.20
JATAKEDDMQNPOQ-UHFFFAOYSA-N
68441
COC1=CC(OC)=C(C(O)=O)C(OC)=C1
C10H12O5
MFCD00016497
benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid
2,4,6-trimethoxybenzoic acid

Specifications

570-02-5
C10H12O5
5 g
benzoic acid, 2,4,6-trimethoxy, 2,4,6-trimethoxy-benzoic acid, pubchem13726, acmc-1awjz, ksc495i9p, 2,4,6-trimethoxy benzoic acid
COC1=CC(OC)=C(C(O)=O)C(OC)=C1
212.20
212.2
2,4,6-Trimethoxybenzoic acid
∽158°C (decomposition)
MFCD00016497
1972294
JATAKEDDMQNPOQ-UHFFFAOYSA-N
2,4,6-trimethoxybenzoic acid
68441
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 209-325-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only