missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide, Thermo Scientific Chemicals

$220.97 - $220.97

Chemical Identifiers

CAS 313981-55-4
Molecular Formula C16H16F3NO2S
Molecular Weight (g/mol) 343.36
MDL Number MFCD00784362
InChI Key SKJJIFRWCCSXGL-UHFFFAOYSA-N
Synonym o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl
PubChem CID 770820
IUPAC Name 2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
SMILES CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ65391MA
View Documents
Thermo Scientific Chemicals
J65391MA
10 mg
Each for $220.97
 
Description

Description

An inactive analog of m-3M3FBS. Used as a negative control.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An inactive analog of m-3M3FBS
  • Used as a negative control
Specifications

Chemical Identifiers

313981-55-4
343.36
SKJJIFRWCCSXGL-UHFFFAOYSA-N
770820
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F
C16H16F3NO2S
MFCD00784362
o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl
2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide

Specifications

313981-55-4
C16H16F3NO2S
10 mg
SKJJIFRWCCSXGL-UHFFFAOYSA-N
2,4,6-trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
770820
Powder
White
MFCD00784362
o-3m3fbs, 2,4,6-trimethyl-n-2-trifluoromethyl phenyl benzenesulfonamide, tocris-1942, benzenesulfonamide,2,4,6-trimethyl-n-2-trifluoromethyl phenyl
CC1=CC(C)=C(C(C)=C1)S(=O)(=O)NC1=CC=CC=C1C(F)(F)F
343.36
343.36
2,4,6-Trimethyl-N-[2-(trifluoromethyl)phenyl]benzenesulfonamide
Videos
Safety and Handling

Safety and Handling

P261-P280a-P305+P351+P338-P304+P340-P405-P501a

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only