Learn More
2,4,6-Trimethylbenzoyl chloride, 98+%, Thermo Scientific Chemicals
$206.36 - $684.82
Chemical Identifiers
CAS | 938-18-1 |
---|---|
Molecular Formula | C10H11ClO |
Molecular Weight (g/mol) | 182.647 |
MDL Number | MFCD00013650 |
InChI Key | UKRQMDIFLKHCRO-UHFFFAOYSA-N |
Synonym | mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride |
PubChem CID | 97038 |
IUPAC Name | 2,4,6-trimethylbenzoyl chloride |
SMILES | CC1=CC(=C(C(=C1)C)C(=O)Cl)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1440709
|
Thermo Scientific Chemicals
A1440709 |
10 g |
N/A
|
|
||||||
AAA1440718
|
Thermo Scientific Chemicals
A1440718 |
50 g |
N/A
|
|
||||||
Description
2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4,6-Trimethylbenzoyl chloride is used, in the presence of pyridine, to protect OH groups as mesitoate esters.
Solubility
Reacts slowly with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C. Air and moisture sensitive. Store under nitrogen. Incompatible with strong oxidizing agent
Chemical Identifiers
938-18-1 | |
182.647 | |
UKRQMDIFLKHCRO-UHFFFAOYSA-N | |
97038 | |
CC1=CC(=C(C(=C1)C)C(=O)Cl)C |
C10H11ClO | |
MFCD00013650 | |
mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride | |
2,4,6-trimethylbenzoyl chloride |
Specifications
938-18-1 | |
143°C to 146°C (60 mmHg) | |
C10H11ClO | |
MFCD00013650 | |
UN3265 | |
Moisture sensitive | |
Reacts slowly with water. | |
CC1=CC(=C(C(=C1)C)C(=O)Cl)C | |
182.647 | |
182.65 | |
2,4,6-Trimethylbenzoyl chloride |
1.1 | |
>110°C (230°F) | |
1.529 | |
10 g | |
776108 | |
mesitoyl chloride, benzoyl chloride, 2,4,6-trimethyl, 2,4,6-trimethylbenzoylchloride, 2,4,6-trimethyl-benzoyl chloride, pubchem13234, acmc-209rno, 2-mesitylenecarbonyl chloride, ksc491c5b, 2,4,6trimethylbenzoyl chloride, 2,4,6-trimethylbenzoyl cloride | |
UKRQMDIFLKHCRO-UHFFFAOYSA-N | |
2,4,6-trimethylbenzoyl chloride | |
97038 | |
≥98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 213-339-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only