missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4,6-Trimethylbenzylamine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 351180050

 View more versions of this product

Catalog No. AC351180050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4, 6-Trimethylbenzylamine
40393-99-5
100.0
Authentic
Glass bottle
1.5360 to 1.538
5 g
mesitylmethanamine, 2,4,6-trimethylbenzylamine, 2,4,6-trimethylphenyl methanamine, 1-mesitylmethanamine, benzenemethanamine, 2,4,6-trimethyl, 1-2,4,6-trimethylphenyl methanamine, 2,4,6-trimethylphenyl methylamine, 2,4,6-trimethylbenzylamine hydrochloride, 1-mesitylmethylamine, pubchem7408
CC1=CC(C)=C(C[NH3+])C(C)=C1
150.24
149.24
Liquid
99%
98.5
Yellow
98.5% min. (GC)
C10H16N
MFCD03411013
12, IV, 2850
DGSRAILDFBJNQI-UHFFFAOYSA-O
(2,4,6-trimethylphenyl)methanamine
458433
99%

Chemical Identifiers

40393-99-5
150.24
DGSRAILDFBJNQI-UHFFFAOYSA-O
458433
C10H16N
MFCD03411013
mesitylmethanamine, 2,4,6-trimethylbenzylamine, 2,4,6-trimethylphenyl methanamine, 1-mesitylmethanamine, benzenemethanamine, 2,4,6-trimethyl, 1-2,4,6-trimethylphenyl methanamine, 2,4,6-trimethylphenyl methylamine, 2,4,6-trimethylbenzylamine hydrochloride, 1-mesitylmethylamine, pubchem7408
CC1=CC(C)=C(C[NH3+])C(C)=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only