Learn More
2,4,6-Trimethylphenol, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 140210100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2, 4, 6-Trimethylphenol | |
98.5 | |
70°C to 73°C | |
220°C | |
98.5% min. (GC) | |
C9H12O | |
MFCD00002235 | |
06, 518 | |
BPRYUXCVCCNUFE-UHFFFAOYSA-N | |
2,4,6-trimethylphenol | |
10698 | |
99% |
527-60-6 | |
100.0 | |
Beige | |
Authentic | |
Glass bottle | |
(CH3)3C6H2OH | |
10 g | |
mesitol, phenol, 2,4,6-trimethyl, mesityl alcohol, 2-hydroxymesitylene, 2,4,6-trimetylofenol, 1,3,5-trimethylphenol, 2,4,6-trimethyl-phenol, 1-hydroxy-2,4,6-trimethylbenzene, 2,4,6-trimethylofenol, unii-fpz32614n6 | |
CC1=CC(=C(C(=C1)C)O)C | |
136.19 | |
136.19 | |
Crystalline Needles |
Chemical Identifiers
527-60-6 | |
136.19 | |
BPRYUXCVCCNUFE-UHFFFAOYSA-N | |
10698 | |
CC1=CC(=C(C(=C1)C)O)C |
C9H12O | |
MFCD00002235 | |
mesitol, phenol, 2,4,6-trimethyl, mesityl alcohol, 2-hydroxymesitylene, 2,4,6-trimetylofenol, 1,3,5-trimethylphenol, 2,4,6-trimethyl-phenol, 1-hydroxy-2,4,6-trimethylbenzene, 2,4,6-trimethylofenol, unii-fpz32614n6 | |
2,4,6-trimethylphenol |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
May cause damage to organs through pr
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 208-419-2
RUO – Research Use Only