missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[4-(Bromomethyl)phenyl]propionic acid, 96%, Thermo Scientific Chemicals

$149.41 - $597.14

Chemical Identifiers

CAS 111128-12-2
Molecular Formula C10H11BrO2
Molecular Weight (g/mol) 243.1
MDL Number MFCD02093445
InChI Key QQXBRVQJMKBAOZ-UHFFFAOYSA-N
Synonym 2-4-bromomethyl phenyl propanoic acid, 2-4-bromomethyl phenyl propionic acid, 2-4-bromomethyl phenylpropionic acid, 2-4-bromomethyl-phenyl-propionic acid, 4-bromomethyl hydratropic acid, 2-4-bromomethyl phenyl propanic acid, 2-p-bromomethyl phenyl propionic acid, 2-4'-bromomethylphenyl propionic acid, 2-4-bromomethyl phenylproplonic acid
PubChem CID 2733978
IUPAC Name 2-[4-(bromomethyl)phenyl]propanoic acid
SMILES CC(C1=CC=C(C=C1)CBr)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6076306
View Documents
Thermo Scientific Chemicals
H6076306
5 g
Each for $149.41
 
AAH6076314
View Documents
Thermo Scientific Chemicals
H6076314
25 g
Each for $597.14
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111128-12-2
243.1
QQXBRVQJMKBAOZ-UHFFFAOYSA-N
2733978
CC(C1=CC=C(C=C1)CBr)C(=O)O
C10H11BrO2
MFCD02093445
2-4-bromomethyl phenyl propanoic acid, 2-4-bromomethyl phenyl propionic acid, 2-4-bromomethyl phenylpropionic acid, 2-4-bromomethyl-phenyl-propionic acid, 4-bromomethyl hydratropic acid, 2-4-bromomethyl phenyl propanic acid, 2-p-bromomethyl phenyl propionic acid, 2-4'-bromomethylphenyl propionic acid, 2-4-bromomethyl phenylproplonic acid
2-[4-(bromomethyl)phenyl]propanoic acid

Specifications

111128-12-2
C10H11BrO2
5 g
2-4-bromomethyl phenyl propanoic acid, 2-4-bromomethyl phenyl propionic acid, 2-4-bromomethyl phenylpropionic acid, 2-4-bromomethyl-phenyl-propionic acid, 4-bromomethyl hydratropic acid, 2-4-bromomethyl phenyl propanic acid, 2-p-bromomethyl phenyl propionic acid, 2-4'-bromomethylphenyl propionic acid, 2-4-bromomethyl phenylproplonic acid
CC(C1=CC=C(C=C1)CBr)C(=O)O
243.1
243.1
2-[4-(Bromomethyl)phenyl]propionic acid
126°C to 130°C
MFCD02093445
UN3261
QQXBRVQJMKBAOZ-UHFFFAOYSA-N
2-[4-(bromomethyl)phenyl]propanoic acid
2733978
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only