missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Bromophenyl)-2-propanol 98.0+%, TCI America™

$200.70 - $600.35

Chemical Identifiers

CAS 2077-19-2
Molecular Formula C9H11BrO
Molecular Weight (g/mol) 215.09
MDL Number MFCD11870096
InChI Key AOGYBHJTXLXRSM-UHFFFAOYSA-N
Synonym 2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, acmc-209fdx, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl-propan-2-ol, 2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, p-bromo-alpha,alpha-dimethylbenzyl alcohol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol
PubChem CID 12681920
IUPAC Name 2-(4-bromophenyl)propan-2-ol
SMILES CC(C)(O)C1=CC=C(Br)C=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B52741G
View Documents
TCI America
B52741G
1 g
Each for $200.70
 
B52745G
View Documents
TCI America
B52745G
5 g
Each for $600.35
 
Description

Description

Specifications

Chemical Identifiers

2077-19-2
215.09
AOGYBHJTXLXRSM-UHFFFAOYSA-N
12681920
CC(C)(O)C1=CC=C(Br)C=C1
C9H11BrO
MFCD11870096
2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, acmc-209fdx, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl-propan-2-ol, 2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, p-bromo-alpha,alpha-dimethylbenzyl alcohol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol
2-(4-bromophenyl)propan-2-ol

Specifications

2077-19-2
White-Yellow
C9H11BrO
1 g
AOGYBHJTXLXRSM-UHFFFAOYSA-N
2-(4-bromophenyl)propan-2-ol
12681920
≥98.0% (GC)
2-(4-Bromophenyl)-2-propanol
46°C
85°C
MFCD11870096
2-4-bromophenyl propan-2-ol, 2-4-bromophenyl-2-propanol, acmc-209fdx, 2-4-bromophenyl propan-2-o1, 2-4-bromophenyl-propan-2-ol, 2-4-bromo-phenyl propan-2-ol, 2-4-bromo-phenyl-propan-2-ol, p-bromo-alpha,alpha-dimethylbenzyl alcohol, 4-bromo-alpha,alpha-dimethylbenzyl alcohol
CC(C)(O)C1=CC=C(Br)C=C1
215.09
215.09
Crystalline Lumps
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'