missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Bromophenyl)ethylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6370606

 View more versions of this product

Catalog No. AAH6370606


Explore available promotions

Description

Description

4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

2-(4-Bromophenyl)ethylamine
Colorless
63°C to 72°C (0.2 mmHg)
C8H11BrN
MFCD00008189
UN2735
Slightly soluble in water.
[NH3+]CCC1=CC=C(Br)C=C1
201.09
200.08
Liquid
73918-56-6
1.290 g/mL
113°C (235°F)
1.574
5 g
4-bromophenethylamine, 2-4-bromophenyl ethylamine, 2-4-bromophenyl ethanamine, p-bromophenethylamine, 2-4-bromo-phenyl-ethylamine, benzeneethanamine, 4-bromo, 2-4-bromophenyl ethan-1-amine, 4-bromophenylethylamine, 4-bromobenzeneethanamine, 1-amino-2-4-bromophenyl ethane
ZSZCXAOQVBEPME-UHFFFAOYSA-O
2-(4-bromophenyl)ethanamine
533915
98%

Chemical Identifiers

73918-56-6
201.09
ZSZCXAOQVBEPME-UHFFFAOYSA-O
533915
C8H11BrN
MFCD00008189
4-bromophenethylamine, 2-4-bromophenyl ethylamine, 2-4-bromophenyl ethanamine, p-bromophenethylamine, 2-4-bromo-phenyl-ethylamine, benzeneethanamine, 4-bromo, 2-4-bromophenyl ethan-1-amine, 4-bromophenylethylamine, 4-bromobenzeneethanamine, 1-amino-2-4-bromophenyl ethane
[NH3+]CCC1=CC=C(Br)C=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 277-636-2

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only