Learn More
2-(4-Bromophenyl)ethylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6370614
Description
4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromophenethylamine was used in the synthesis of pyrazinoisoquinoline derivatives and N-2-(4-bromophenyl)ethyl chloroacetamide. It was also used in the synthesis of alkyl arylamino sufides employing elemental sulfur and various halides.
Solubility
Slightly soluble in water.
Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
2-(4-Bromophenyl)ethylamine | |
Colorless | |
63°C to 72°C (0.2 mmHg) | |
C8H11BrN | |
MFCD00008189 | |
UN2735 | |
Slightly soluble in water. | |
[NH3+]CCC1=CC=C(Br)C=C1 | |
201.09 | |
200.08 | |
Liquid |
73918-56-6 | |
1.290 g/mL | |
113°C (235°F) | |
1.574 | |
25 g | |
4-bromophenethylamine, 2-4-bromophenyl ethylamine, 2-4-bromophenyl ethanamine, p-bromophenethylamine, 2-4-bromo-phenyl-ethylamine, benzeneethanamine, 4-bromo, 2-4-bromophenyl ethan-1-amine, 4-bromophenylethylamine, 4-bromobenzeneethanamine, 1-amino-2-4-bromophenyl ethane | |
ZSZCXAOQVBEPME-UHFFFAOYSA-O | |
2-(4-bromophenyl)ethanamine | |
533915 | |
98% |
Chemical Identifiers
73918-56-6 | |
201.09 | |
ZSZCXAOQVBEPME-UHFFFAOYSA-O | |
533915 |
C8H11BrN | |
MFCD00008189 | |
4-bromophenethylamine, 2-4-bromophenyl ethylamine, 2-4-bromophenyl ethanamine, p-bromophenethylamine, 2-4-bromo-phenyl-ethylamine, benzeneethanamine, 4-bromo, 2-4-bromophenyl ethan-1-amine, 4-bromophenylethylamine, 4-bromobenzeneethanamine, 1-amino-2-4-bromophenyl ethane | |
[NH3+]CCC1=CC=C(Br)C=C1 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 277-636-2
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only