missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)furan, ≥97%, Thermo Scientific™

Chemical Identifiers

CAS 14297-34-8
Molecular Formula C10H7BrO
Molecular Weight (g/mol) 223.069
MDL Number MFCD07775614
InChI Key FBIKSVALROQWIY-UHFFFAOYSA-N
Synonym 2-4-bromophenyl furan, 2-4-bromophenyl-furan, pubchem2039, 2 4-bromophenyl furan, 2-p-bromophenyl furan, 4-bromo-2-furyl benzene, furan, 2-4-bromophenyl
PubChem CID 11276114
IUPAC Name 2-(4-bromophenyl)furan
SMILES C1=COC(=C1)C2=CC=C(C=C2)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC34410CB
SDS View Documents
Thermo Scientific Chemicals
CC34410CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34410DA
SDS View Documents
Thermo Scientific Chemicals
CC34410DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

14297-34-8
223.069
FBIKSVALROQWIY-UHFFFAOYSA-N
11276114
C1=COC(=C1)C2=CC=C(C=C2)Br
C10H7BrO
MFCD07775614
2-4-bromophenyl furan, 2-4-bromophenyl-furan, pubchem2039, 2 4-bromophenyl furan, 2-p-bromophenyl furan, 4-bromo-2-furyl benzene, furan, 2-4-bromophenyl
2-(4-bromophenyl)furan

Specifications

14297-34-8
100.0
C10H7BrO
250mg
FBIKSVALROQWIY-UHFFFAOYSA-N
2-(4-bromophenyl)furan
11276114
97%
96.5
97%
MFCD07775614
2-4-bromophenyl furan, 2-4-bromophenyl-furan, pubchem2039, 2 4-bromophenyl furan, 2-p-bromophenyl furan, 4-bromo-2-furyl benzene, furan, 2-4-bromophenyl
C1=COC(=C1)C2=CC=C(C=C2)Br
223.069
223.07
2-(4-Bromophenyl)furan
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'