missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Chlorophenyl)-4,5-diphenylimidazole 98.0+%, TCI America™
Supplier: TCI America C2990200MG
Specifications
| 2-(4-Chlorophenyl)-4,5-diphenylimidazole | |
| 265°C | |
| C21H15ClN2 | |
| 200 mg | |
| C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=C(C=C3)Cl)C4=CC=CC=C4 | |
| 330.815 | |
| 330.82 | |
| Crystalline Powder |
| 5496-32-2 | |
| White-Yellow | |
| MFCD00030329 | |
| NJXBHEMXPUMJRO-UHFFFAOYSA-N | |
| 2-(4-chlorophenyl)-4,5-diphenyl-1H-imidazole | |
| 794727 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 5496-32-2 | |
| 330.815 | |
| NJXBHEMXPUMJRO-UHFFFAOYSA-N | |
| 2-(4-chlorophenyl)-4,5-diphenyl-1H-imidazole |
| C21H15ClN2 | |
| MFCD00030329 | |
| 794727 | |
| C1=CC=C(C=C1)C2=C(N=C(N2)C3=CC=C(C=C3)Cl)C4=CC=CC=C4 |
Safety and Handling
TSCA : No