Learn More
2-(4-Chlorophenyl)ethylamine, 97%, Thermo Scientific Chemicals
$62.84 - $562.47
Chemical Identifiers
CAS | 156-41-2 |
---|---|
Molecular Formula | C8H11ClN |
Molecular Weight (g/mol) | 156.63 |
MDL Number | MFCD00008191 |
InChI Key | SRXFXCKTIGELTI-UHFFFAOYSA-O |
Synonym | 4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine |
PubChem CID | 67430 |
IUPAC Name | 2-(4-chlorophenyl)ethanamine |
SMILES | [NH3+]CCC1=CC=C(Cl)C=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH6308106
|
Thermo Scientific Chemicals
H6308106 |
5 g |
Each for $62.84
|
|
||||||
AAH6308114
|
Thermo Scientific Chemicals
H6308114 |
25 g |
Each for $217.32
|
|
||||||
AAH6308122
|
Thermo Scientific Chemicals
H6308122 |
100 g |
Each for $562.47
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
156-41-2 | |
156.63 | |
SRXFXCKTIGELTI-UHFFFAOYSA-O | |
67430 | |
[NH3+]CCC1=CC=C(Cl)C=C1 |
C8H11ClN | |
MFCD00008191 | |
4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine | |
2-(4-chlorophenyl)ethanamine |
Specifications
156-41-2 | |
60°C to 65°C (0.1 mmHg) | |
C8H11ClN | |
5 g | |
4-chlorophenethylamine, 2-4-chlorophenyl ethylamine, 2-4-chlorophenyl ethanamine, 2-p-chlorophenyl ethylamine, p-chlorophenethylamine, 2-4-chlorophenyl ethan-1-amine, 4-chlorobenzeneethanamine, 2-4-chloro-phenyl-ethylamine, benzeneethanamine, 4-chloro, beta-p-chlorophenyl ethylamine | |
[NH3+]CCC1=CC=C(Cl)C=C1 | |
156.63 | |
155.62 | |
Liquid |
1.120 g/mL | |
106°C (223°F) | |
MFCD00008191 | |
508247 | |
SRXFXCKTIGELTI-UHFFFAOYSA-O | |
2-(4-chlorophenyl)ethanamine | |
67430 | |
97% | |
2-(4-Chlorophenyl)ethylamine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
EINECSNumber : 205-853-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only