missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Chlorophenyl)malondialdehyde, 95%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 425200025

Catalog No. AC425200025


Explore available promotions

Description

Description

 

Specifications

Specifications

2-(4-Chlorophenyl)malondialdehyde
205676-17-1
100.0
Beige or White to Yellow
94% min. (HPLC)
C9H7ClO2
2.5 g
Solubility in water: negligible
O\C=C(/C=O)C1=CC=C(Cl)C=C1
182.60
182.61
Crystalline Powder
95%
94.0
137°C to 143°C
Authentic
Glass bottle
MFCD00216505
2-4-chlorophenyl malonaldehyde, 2-4-chlorophenyl propanedial, 2-4-chlorophenyl malondialdehyde, 4-chlorophenyl propane-1,3-dial, 2-4-chlorophenyl malon-dialdehyde, 2-4-chlorophenyl propane-1,3-dione
BPCQPZCKKNFWCF-VMPITWQZSA-N
2-(4-chlorophenyl)propanedial
3584386
95%

Chemical Identifiers

205676-17-1
182.60
BPCQPZCKKNFWCF-VMPITWQZSA-N
3584386
O\C=C(/C=O)C1=CC=C(Cl)C=C1
C9H7ClO2
MFCD00216505
2-4-chlorophenyl malonaldehyde, 2-4-chlorophenyl propanedial, 2-4-chlorophenyl malondialdehyde, 4-chlorophenyl propane-1,3-dial, 2-4-chlorophenyl malon-dialdehyde, 2-4-chlorophenyl propane-1,3-dione
2-(4-chlorophenyl)propanedial
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only