missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Diaminopyrimidine 98.0+%, TCI America™

$371.30 - $1171.17

Chemical Identifiers

CAS 156-81-0
Molecular Formula C4H6N4
Molecular Weight (g/mol) 110.12
MDL Number MFCD00038023
InChI Key YAAWASYJIRZXSZ-UHFFFAOYSA-N
Synonym 2,4-diaminopyrimidine, 2,4-pyrimidinediamine, 2,6-diaminopyrimidine, pyrimidinediamine, diaminopyrimidine, pyrimidine, 2,4-diamino, 2,4-diamino-pyrimidine, pyrimidine-2,4-diyldiamine, 2,4-pyrimidinediamine 9ci, 4-amino-2-pyrimidineamine
PubChem CID 67431
ChEBI CHEBI:43745
IUPAC Name pyrimidine-2,4-diamine
SMILES C1=CN=C(N=C1N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D21205G
View Documents
TCI America
D21205G
5 g
Each for $371.30
 
D212025G
View Documents
TCI America
D212025G
25 g
Each for $1,171.17
 
Description

Description

Specifications

Chemical Identifiers

156-81-0
110.12
YAAWASYJIRZXSZ-UHFFFAOYSA-N
67431
pyrimidine-2,4-diamine
C4H6N4
MFCD00038023
2,4-diaminopyrimidine, 2,4-pyrimidinediamine, 2,6-diaminopyrimidine, pyrimidinediamine, diaminopyrimidine, pyrimidine, 2,4-diamino, 2,4-diamino-pyrimidine, pyrimidine-2,4-diyldiamine, 2,4-pyrimidinediamine 9ci, 4-amino-2-pyrimidineamine
CHEBI:43745
C1=CN=C(N=C1N)N

Specifications

156-81-0
White-Yellow
MFCD00038023
2,4-diaminopyrimidine, 2,4-pyrimidinediamine, 2,6-diaminopyrimidine, pyrimidinediamine, diaminopyrimidine, pyrimidine, 2,4-diamino, 2,4-diamino-pyrimidine, pyrimidine-2,4-diyldiamine, 2,4-pyrimidinediamine 9ci, 4-amino-2-pyrimidineamine
C1=CN=C(N=C1N)N
110.12
CHEBI:43745
≥98.0% (GC,T)
2,4-Diaminopyrimidine
149°C
C4H6N4
5 g
YAAWASYJIRZXSZ-UHFFFAOYSA-N
pyrimidine-2,4-diamine
67431
110.12
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'