missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dibromo-1-nitrobenzene 98.0+%, TCI America™
$42.87 - $108.11
Chemical Identifiers
| CAS | 51686-78-3 |
|---|---|
| Molecular Formula | C6H3Br2NO2 |
| Molecular Weight (g/mol) | 280.903 |
| MDL Number | MFCD02660416 |
| InChI Key | DXRVYZGVVFZCFP-UHFFFAOYSA-N |
| Synonym | 2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene |
| PubChem CID | 12315646 |
| IUPAC Name | 2,4-dibromo-1-nitrobenzene |
| SMILES | C1=CC(=C(C=C1Br)Br)[N+](=O)[O-] |
Chemical Identifiers
| 51686-78-3 | |
| 280.903 | |
| DXRVYZGVVFZCFP-UHFFFAOYSA-N | |
| 12315646 | |
| C1=CC(=C(C=C1Br)Br)[N+](=O)[O-] |
| C6H3Br2NO2 | |
| MFCD02660416 | |
| 2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene | |
| 2,4-dibromo-1-nitrobenzene |
Specifications
| 51686-78-3 | |
| Yellow | |
| MFCD02660416 | |
| 2,4-dibromonitrobenzene, benzene, 2,4-dibromo-1-nitro, 2,4-dibromo-1-nitro-benzene, 1,3-dibromo-4-nitrobenzene, acmc-209kve, 2,4-dibromo-nitrobenzene, ksc594m9r, benzene,2,4-dibromo-1-nitro, 2,4-bis bromanyl-1-nitro-benzene, 2,4-dibromo-1-nitrobenzene;,3-dibromo-4-nitrobenzene | |
| C1=CC(=C(C=C1Br)Br)[N+](=O)[O-] | |
| 280.903 | |
| 280.90 | |
| Crystalline Powder |
| 61°C | |
| C6H3Br2NO2 | |
| 1 g | |
| DXRVYZGVVFZCFP-UHFFFAOYSA-N | |
| 2,4-dibromo-1-nitrobenzene | |
| 12315646 | |
| ≥98.0% (GC) | |
| 2,4-Dibromo-1-nitrobenzene |
Safety and Handling
TSCA : No