Learn More
2,4-Dibromo-3-pentanone, mixture of stereoisomers, 97%, Thermo Scientific Chemicals
$235.58 - $880.22
Chemical Identifiers
CAS | 815-60-1 |
---|---|
Molecular Formula | C5H8Br2O |
Molecular Weight (g/mol) | 243.926 |
MDL Number | MFCD00015719 |
InChI Key | UOPIOAUZQKSZRO-UHFFFAOYSA-N |
Synonym | 2,4-dibromo-3-pentanone, 2,4-dibromo-pentan-3-one, 3-pentanone,2,4-dibromo, 2,4-bis bromanyl pentan-3-one, 2,4-dibromo-3-pentanone, mixture of stereoisomers |
PubChem CID | 11791395 |
IUPAC Name | 2,4-dibromopentan-3-one |
SMILES | CC(C(=O)C(C)Br)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1668009
|
Thermo Scientific Chemicals
A1668009 |
10 g |
Each for $235.58
|
|
|||||
AAA1668018
|
Thermo Scientific Chemicals
A1668018 |
50 g |
Each for $880.22
|
|
|||||
Description
2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dibromo-2,4-dimethyl-3-pentanone may be used in the synthesis of 2-dimethylamino-4-methylene-1,3-dioxolanes, via debromination using zinc-copper couple in dimethylformamide and dimethylacetamide.
Solubility
Slightly soluble in water(50 g/L (20°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, reducing agents, bases.
Chemical Identifiers
815-60-1 | |
243.926 | |
UOPIOAUZQKSZRO-UHFFFAOYSA-N | |
11791395 | |
CC(C(=O)C(C)Br)Br |
C5H8Br2O | |
MFCD00015719 | |
2,4-dibromo-3-pentanone, 2,4-dibromo-pentan-3-one, 3-pentanone,2,4-dibromo, 2,4-bis bromanyl pentan-3-one, 2,4-dibromo-3-pentanone, mixture of stereoisomers | |
2,4-dibromopentan-3-one |
Specifications
815-60-1 | |
Pungent | |
MFCD00015719 | |
UN3265 | |
Slightly soluble in water(50g/L (20°C). | |
CC(C(=O)C(C)Br)Br | |
243.926 | |
243.91 | |
2,4-Dibromo-3-pentanone, mixture of stereoisomers |
67°C to 82°C (10 mmHg) | |
C5H8Br2O | |
10 g | |
2,4-dibromo-3-pentanone, 2,4-dibromo-pentan-3-one, 3-pentanone,2,4-dibromo, 2,4-bis bromanyl pentan-3-one, 2,4-dibromo-3-pentanone, mixture of stereoisomers | |
UOPIOAUZQKSZRO-UHFFFAOYSA-N | |
2,4-dibromopentan-3-one | |
11791395 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only