missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dichloro-5-methylpyridine, 96%, Thermo Scientific™
$130.36 - $130.36
Chemical Identifiers
CAS | 56961-78-5 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.01 |
MDL Number | MFCD08704813 |
InChI Key | DUPJNQNLAKJWKU-UHFFFAOYSA-N |
Synonym | 2,4-dichloro-5-picoline, pyridine, 2,4-dichloro-5-methyl, 2,4-dichloro-5-methyl-pyridine, pubchem5464, acmc-209lv3, ksc495i9n, 2,4-dichloro-5-methyl pyridine, 2,4-bis chloranyl-5-methyl-pyridine |
PubChem CID | 12643718 |
IUPAC Name | 2,4-dichloro-5-methylpyridine |
SMILES | CC1=CN=C(Cl)C=C1Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6163403
|
Thermo Scientific Chemicals
H6163403 |
1 g |
Each for $130.36
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2,4-Dichloro-5-methylpyridine is used as pharmaceutical intermediates.
Solubility
Slightly soluble in water.
Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
Chemical Identifiers
56961-78-5 | |
162.01 | |
DUPJNQNLAKJWKU-UHFFFAOYSA-N | |
12643718 | |
CC1=CN=C(Cl)C=C1Cl |
C6H5Cl2N | |
MFCD08704813 | |
2,4-dichloro-5-picoline, pyridine, 2,4-dichloro-5-methyl, 2,4-dichloro-5-methyl-pyridine, pubchem5464, acmc-209lv3, ksc495i9n, 2,4-dichloro-5-methyl pyridine, 2,4-bis chloranyl-5-methyl-pyridine | |
2,4-dichloro-5-methylpyridine |
Specifications
56961-78-5 | |
C6H5Cl2N | |
1 g | |
Slightly soluble in water. | |
CC1=CN=C(Cl)C=C1Cl | |
162.01 | |
162.02 | |
Liquid |
Colorless | |
MFCD08704813 | |
2,4-dichloro-5-picoline, pyridine, 2,4-dichloro-5-methyl, 2,4-dichloro-5-methyl-pyridine, pubchem5464, acmc-209lv3, ksc495i9n, 2,4-dichloro-5-methyl pyridine, 2,4-bis chloranyl-5-methyl-pyridine | |
DUPJNQNLAKJWKU-UHFFFAOYSA-N | |
2,4-dichloro-5-methylpyridine | |
12643718 | |
96% | |
2,4-Dichloro-5-methylpyridine |
Safety and Handling
GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
TSCA : No
Recommended Storage : Ambient temperatures