Learn More
2,4-Dichloro-6-methylpyrimidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1036314
Description
2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.
Solubility
Soluble in Chloroform, Ether, Ethyl Acetate and Toluene. Insoluble in water.
Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
2,4-Dichloro-6-methylpyrimidine | |
44°C to 47°C | |
>110°C (230°F) | |
MFCD00006064 | |
UN1759 | |
pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine | |
BTLKROSJMNFSQZ-UHFFFAOYSA-N | |
2,4-dichloro-6-methylpyrimidine | |
79471 | |
98% |
5424-21-5 | |
219°C | |
C5H4Cl2N2 | |
25 g | |
114295 | |
Soluble in Chloroform,Ether,Ethyl Acetate and Toluene. Insoluble in water. | |
CC1=CC(=NC(=N1)Cl)Cl | |
163.001 | |
163.01 |
Chemical Identifiers
5424-21-5 | |
163.001 | |
BTLKROSJMNFSQZ-UHFFFAOYSA-N | |
79471 | |
CC1=CC(=NC(=N1)Cl)Cl |
C5H4Cl2N2 | |
MFCD00006064 | |
pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine | |
2,4-dichloro-6-methylpyrimidine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 226-563-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only