missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichloro-6-methylpyrimidine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1036322

 View more versions of this product

Catalog No. AAA1036322


Explore available promotions

Description

Description

2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dichloro-6-methylpyrimidine is used in the synthesis of (2-chloro-6-methyl-pyrimidin-4-yl)-(2,3-dihydro-benzothiazol-6-yl)-amine.

Solubility
Soluble in Chloroform, Ether, Ethyl Acetate and Toluene. Insoluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2,4-Dichloro-6-methylpyrimidine
44°C to 47°C
>110°C (230°F)
MFCD00006064
UN1759
pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine
BTLKROSJMNFSQZ-UHFFFAOYSA-N
2,4-dichloro-6-methylpyrimidine
79471
98%
5424-21-5
219°C
C5H4Cl2N2
100 g
114295
Soluble in Chloroform,Ether,Ethyl Acetate and Toluene. Insoluble in water.
CC1=CC(=NC(=N1)Cl)Cl
163.001
163.01

Chemical Identifiers

5424-21-5
163.001
BTLKROSJMNFSQZ-UHFFFAOYSA-N
79471
CC1=CC(=NC(=N1)Cl)Cl
C5H4Cl2N2
MFCD00006064
pyrimidine, 2,4-dichloro-6-methyl, 2,6-dichloro-4-methylpyrimidine, 2,4-dichloro-6-methyl-pyrimidine, 2,4-dichloro-6-methyl pyrimidine, 6-methyl-2,4-dichloropyrimidine, 2,4-dichlor-6-methylpyrimidin, pubchem6911, acmc-1b11m, ksc495i8r, 2,4-dichloro6-methylpyrimidine
2,4-dichloro-6-methylpyrimidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

EINECSNumber : 226-563-4

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only