missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dichloroacetophenone, 97%, Thermo Scientific™

Chemical Identifiers

CAS 2234-16-4
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000581
InChI Key XMCRWEBERCXJCH-UHFFFAOYSA-N
Synonym 2',4'-dichloroacetophenone, 1-2,4-dichlorophenyl ethanone, 2,4-dichloroacetophenone, ethanone, 1-2,4-dichlorophenyl, 1-2,4-dichlorophenyl ethan-1-one, p-chloro-2-chloroacetophenone, acetophenone, 2',4'-dichloro, unii-t7f2608h5h, 2',4'-dichloro phenyl ethanone, 1-2,4-dichloro-phenyl-ethanone
PubChem CID 16693
IUPAC Name 1-(2,4-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC302430250
SDS View Documents
Thermo Scientific Chemicals
302430250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC302432500
SDS View Documents
Thermo Scientific Chemicals
302432500
250g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

2234-16-4
189.04
XMCRWEBERCXJCH-UHFFFAOYSA-N
16693
CC(=O)C1=C(C=C(C=C1)Cl)Cl
C8H6Cl2O
MFCD00000581
2',4'-dichloroacetophenone, 1-2,4-dichlorophenyl ethanone, 2,4-dichloroacetophenone, ethanone, 1-2,4-dichlorophenyl, 1-2,4-dichlorophenyl ethan-1-one, p-chloro-2-chloroacetophenone, acetophenone, 2',4'-dichloro, unii-t7f2608h5h, 2',4'-dichloro phenyl ethanone, 1-2,4-dichloro-phenyl-ethanone
1-(2,4-dichlorophenyl)ethanone

Specifications

2234-16-4
100.0
242.0°C
Authentic
Glass bottle
1.5635
MFCD00000581
07, II, 219
2',4'-dichloroacetophenone, 1-2,4-dichlorophenyl ethanone, 2,4-dichloroacetophenone, ethanone, 1-2,4-dichlorophenyl, 1-2,4-dichlorophenyl ethan-1-one, p-chloro-2-chloroacetophenone, acetophenone, 2',4'-dichloro, unii-t7f2608h5h, 2',4'-dichloro phenyl ethanone, 1-2,4-dichloro-phenyl-ethanone
XMCRWEBERCXJCH-UHFFFAOYSA-N
1-(2,4-dichlorophenyl)ethanone
16693
97%
96.0
1.3200g/mL
131°C
96% min. (GC)
C8H6Cl2O
Cl2C6H3COCH3
25g
1.32
Solubility in water: ca. 200mg/l in water
CC(=O)C1=C(C=C(C=C1)Cl)Cl
189.04
189.04
2', 4'-Dichloroacetophenone, 98.5%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-780-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only