Learn More
2,4-Dichloroaniline, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1051822
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2,4-Dichloroaniline | |
59°C to 63°C | |
242°C | |
C6H5Cl2N | |
100 g | |
386422 | |
KQCMTOWTPBNWDB-UHFFFAOYSA-N | |
2,4-dichloroaniline | |
11123 | |
162.02 |
554-00-7 | |
1.567 | |
149°C (300°F) | |
MFCD00007661 | |
UN3442 | |
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german | |
C1=CC(=C(C=C1Cl)Cl)N | |
162.013 | |
CHEBI:46635 | |
99% |
Chemical Identifiers
554-00-7 | |
162.013 | |
KQCMTOWTPBNWDB-UHFFFAOYSA-N | |
11123 | |
2,4-dichloroaniline |
C6H5Cl2N | |
MFCD00007661 | |
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german | |
CHEBI:46635 | |
C1=CC(=C(C=C1Cl)Cl)N |
Safety and Handling
GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c
H301+H311+H331-H373-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
EINECSNumber : 209-057-8
RTECSNumber : BX2600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only