missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dichloronitrobenzene 99.0+%, TCI America™
Supplier: TCI America D098225G
Specifications
2,4-Dichloronitrobenzene | |
White-Yellow | |
C6H3Cl2NO2 | |
25 g | |
2,4-dichloronitrobenzene, 1,3-dichloro-4-nitrobenzene, benzene, 2,4-dichloro-1-nitro, unii-85p1a896rr, ccris 3095, benzene, 1,3-dichloro-4-nitro, nitro-m-dichlorobenzene, pubchem15890, 2,4-dichloronitrobenzen, 4-nitro-m-dichlorobenzene | |
C1=CC(=C(C=C1Cl)Cl)[N+](=O)[O-] | |
191.995 | |
192.00 | |
Crystalline Powder |
611-06-3 | |
259°C | |
MFCD00007071 | |
2811 | |
QUIMTLZDMCNYGY-UHFFFAOYSA-N | |
2,4-dichloro-1-nitrobenzene | |
11899 | |
≥99.0% (GC) |
Chemical Identifiers
611-06-3 | |
191.995 | |
QUIMTLZDMCNYGY-UHFFFAOYSA-N | |
11899 | |
C1=CC(=C(C=C1Cl)Cl)[N+](=O)[O-] |
C6H3Cl2NO2 | |
MFCD00007071 | |
2,4-dichloronitrobenzene, 1,3-dichloro-4-nitrobenzene, benzene, 2,4-dichloro-1-nitro, unii-85p1a896rr, ccris 3095, benzene, 1,3-dichloro-4-nitro, nitro-m-dichlorobenzene, pubchem15890, 2,4-dichloronitrobenzen, 4-nitro-m-dichlorobenzene | |
2,4-dichloro-1-nitrobenzene |
Safety and Handling
EINECSNumber : (3)-0455
RTECSNumber : CZ5420000
TSCA : Yes