missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorophenol, 99%, Thermo Scientific Chemicals

$37.22 - $144.60

Chemical Identifiers

CAS 120-83-2
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002169
InChI Key HFZWRUODUSTPEG-UHFFFAOYSA-N
Synonym 4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
PubChem CID 8449
ChEBI CHEBI:16738
IUPAC Name 2,4-dichlorophenol
SMILES OC1=CC=C(Cl)C=C1Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC147720050
SDS View Documents
Thermo Scientific Chemicals
147720050
5 g Glass bottle
Each for $37.22
 
AC147721000
SDS View Documents
Thermo Scientific Chemicals
147721000
100 g Glass bottle
Each for $39.89
 
AC147725000
SDS View Documents
Thermo Scientific Chemicals
147725000
500 g Glass bottle
Each for $144.60
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

120-83-2
163.00
HFZWRUODUSTPEG-UHFFFAOYSA-N
8449
2,4-dichlorophenol
C6H4Cl2O
MFCD00002169
4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
CHEBI:16738
OC1=CC=C(Cl)C=C1Cl

Specifications

120-83-2
100.0
White to Beige
113°C
98.5% min. (GC)
C6H4Cl2O
MFCD00002169
06, 189
4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
HFZWRUODUSTPEG-UHFFFAOYSA-N
2,4-dichlorophenol
8449
163
Crystalline Solid
98.5
41°C to 44°C
209°C to 210°C
Authentic
Glass bottle
Cl2C6H3OH
5 g
15, 3082
Solubility in water: 4.5g/L in water (20°C). Other solubilities: soluble in carbon tetrachloride,diethyl ether,,benzene,chloroform,ethanol,aromatic solvents,,acetone and sodium hydroxide
OC1=CC=C(Cl)C=C1Cl
163.00
CHEBI:16738
99%
2, 4-Dichlorophenol, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 204-429-6

RTECSNumber : SK8575000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only