missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dichlorophenylacetic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1179106

 View more versions of this product

Catalog No. AAA1179106


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dichlorophenylacetic acid
128°C to 130°C
C8H6Cl2O2
5 g
2,4-dichlorophenylacetic acid, 2-2,4-dichlorophenyl acetic acid, benzeneacetic acid, 2,4-dichloro, 2,4-dichloro-phenyl-acetic acid, 2,4-dichlorophenyl acetic acid, 2 ,4-dichlorophenylacetic acid, zlchem 477, pubchem15026, acmc-209f0x
C1=CC(=C(C=C1Cl)Cl)CC(=O)O
205.034
205.04
19719-28-9
Odorless
MFCD00004318
1948767
GXMWLJKTGBZMBH-UHFFFAOYSA-N
2-(2,4-dichlorophenyl)acetic acid
88209
≥98%

Chemical Identifiers

19719-28-9
205.034
GXMWLJKTGBZMBH-UHFFFAOYSA-N
2-(2,4-dichlorophenyl)acetic acid
C8H6Cl2O2
MFCD00004318
88209
C1=CC(=C(C=C1Cl)Cl)CC(=O)O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 243-248-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only