missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Difluoro-5-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3261714

 View more versions of this product

Catalog No. AAH3261714


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Difluoro-5-nitrobenzoic acid
125°C to 133°C
MFCD04115713
2,4-difluoro-5-nitrobenzoic acid, benzoic acid, 2,4-difluoro-5-nitro, benzoic acid,2,4-difluoro-5-nitro, 2,4-difluoro-5-nitro-benzoic acid, acmc-209d9v, ksc527s6t, 2,4-difluoro-5-nitrobenzoicacid, buttpark 156\01-93
[O-]C(=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O
202.09
203.1
153775-33-8
C7H2F2NO4
25 g
MDFSGDMPHMKKGB-UHFFFAOYSA-M
2,4-difluoro-5-nitrobenzoate
2782822
97%

Chemical Identifiers

153775-33-8
202.09
MDFSGDMPHMKKGB-UHFFFAOYSA-M
2,4-difluoro-5-nitrobenzoate
C7H2F2NO4
MFCD04115713
2782822
[O-]C(=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only